Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930121.1 | - | 1 - 2 | 87 | 120 | |
BX930576.2 | - | 1 - 2 | 87 | 120 | ||
BX930906.1 | - | 1 - 2 | 100 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |