Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277137.1 | NP_001264066.1 | 1 - 2 | 113 | 68 | |
GenBank mRNA | BX933453.2 | - | 1 - 2 | 123 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |