Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277137.1 | NP_001264066.1 | 4 - 5 | 703 | 117 | |
NM_001277138.1 | NP_001264067.1 | 6 - 7 | 795 | 117 | ||
GenBank mRNA | BX933453.2 | - | 4 - 5 | 707 | 117 | |
BX935073.1 | - | 6 - 7 | 795 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |