Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145016.1 | NP_001138488.1 | 7 - 8 | 1088 | 70 | |
XM_015296352.2 | XP_015151838.1 | 7 - 8 | 1338 | 70 | ||
XM_015296353.2 | XP_015151839.1 | 7 - 8 | 1338 | 70 | ||
XM_015296354.2 | XP_015151840.1 | 7 - 8 | 1169 | 70 | ||
GenBank mRNA | BX934665.2 | - | 4 - 5 | 471 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |