Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418224.6 | XP_418224.2 | 5 - 6 | 525 | 133 | |
GenBank mRNA | BX929663.1 | - | 5 - 6 | 509 | 133 | |
BX935423.2 | - | 5 - 6 | 512 | 133 | ||
BX935442.2 | - | 5 - 6 | 500 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |