Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278050.3 | NP_001264979.1 | 1 - 2 | 485 | 112 | |
GenBank mRNA | BX950715.2 | - | 1 - 2 | 485 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |