Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936311.3 | XP_004936368.1 | 6 - 7 | 1420 | 141 | |
XM_004936312.2 | XP_004936369.2 | 5 - 6 | 719 | 141 | ||
XM_015287784.2 | XP_015143270.1 | 5 - 6 | 829 | 141 | ||
XM_015287785.2 | XP_015143271.1 | 6 - 7 | 1282 | 141 | ||
XM_015287786.2 | XP_015143272.1 | 7 - 8 | 1324 | 141 | ||
XM_015287787.2 | XP_015143273.1 | 2 - 3 | 643 | 141 | ||
XM_025150906.1 | XP_025006674.1 | 5 - 6 | 718 | 141 | ||
XM_025150907.1 | XP_025006675.1 | 2 - 3 | 464 | 141 | ||
XM_025150908.1 | XP_025006676.1 | 5 - 6 | 600 | 141 | ||
XM_025150909.1 | XP_025006677.1 | 4 - 5 | 588 | 141 | ||
XM_421367.6 | XP_421367.3 | 5 - 6 | 831 | 141 | ||
GenBank mRNA | CR353166.1 | - | 1 - 2 | 283 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |