Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416415.6 | XP_416415.5 | 7 - 8 | 1562 | 91 | |
GenBank mRNA | CR353321.1 | - | 7 - 8 | 877 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |