Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199396.1 | NP_001186325.1 | 19 - 20 | 2154 | 82 | |
XM_015283472.2 | XP_015138958.1 | 19 - 20 | 2258 | 82 | ||
XM_025148594.1 | XP_025004362.1 | 17 - 18 | 2065 | 82 | ||
XM_025148595.1 | XP_025004363.1 | 16 - 17 | 1786 | 82 | ||
GenBank mRNA | CR353555.1 | - | 10 - 11 | 1058 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |