Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199525.1 | NP_001186454.1 | 3 - 4 | 94 | ||
GenBank mRNA | CR354049.1 | - | 3 - 4 | 529 | 94 | |
CR354102.1 | - | 3 - 4 | 616 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |