Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143969.1 | NP_001137441.1 | 2 - 3 | 173 | 146 | |
GenBank mRNA | CR385805.1 | - | 2 - 3 | 173 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |