Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039263.2 | NP_001034352.2 | 2 - 3 | 208 | 84 | |
XM_015294144.2 | XP_015149630.1 | 2 - 3 | 201 | 84 | ||
GenBank mRNA | CR387393.1 | - | 2 - 3 | 195 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |