Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151998.1 | XP_025007766.1 | 1 - 2 | 13296 | 101 | |
GenBank mRNA | AY753292.1 | - | 1 - 2 | 146 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |