Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199574.1 | NP_001186503.1 | 5 - 6 | 862 | 72 | |
XM_015286081.2 | XP_015141567.1 | 5 - 6 | 907 | 72 | ||
XM_015286082.2 | XP_015141568.1 | 5 - 6 | 772 | 72 | ||
XM_015286083.2 | XP_015141569.1 | 5 - 6 | 852 | 72 | ||
GenBank mRNA | BX930104.2 | - | 4 - 5 | 752 | 72 | |
BX934774.2 | - | 5 - 6 | 812 | 72 | ||
BX950333.1 | - | 5 - 6 | 862 | 72 | ||
CR406727.1 | - | 4 - 5 | 267 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |