Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420875.6 | XP_420875.5 | 1 - 2 | 102 | 84 | |
GenBank mRNA | BX934322.2 | - | 1 - 2 | 52 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |