Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199517.1 | NP_001186446.1 | 1 - 2 | 70 | 125 | |
GenBank mRNA | BX950529.2 | - | 1 - 2 | 58 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |