Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025153495.1 | XP_025009263.1 | 2 - 3 | 383 | 107 | |
XM_025153499.1 | XP_025009267.1 | 2 - 3 | 383 | 107 | ||
XM_025153501.1 | XP_025009269.1 | 2 - 3 | 558 | 107 | ||
GenBank mRNA | CR387037.1 | - | 2 - 3 | 387 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |