Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422723.6 | XP_422723.3 | 4 - 5 | 715 | 83 | |
GenBank mRNA | CR390246.1 | - | 3 - 4 | 230 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |