Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941658.3 | XP_004941715.1 | 13 - 14 | 4156 | 67 | |
XM_004941659.3 | XP_004941716.1 | 12 - 13 | 4050 | 67 | ||
XM_025151266.1 | XP_025007034.1 | 13 - 14 | 3648 | 67 | ||
XM_025151267.1 | XP_025007035.1 | 14 - 15 | 3694 | 67 | ||
XM_025151268.1 | XP_025007036.1 | 12 - 13 | 3542 | 67 | ||
XM_025151269.1 | XP_025007037.1 | 12 - 13 | 3382 | 67 | ||
GenBank mRNA | CR391145.1 | - | 1 - 2 | 76 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |