Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276340.2 | XP_015131826.1 | 23 - 24 | 4841 | 72 | |
XM_015276342.2 | XP_015131828.1 | 23 - 24 | 4799 | 72 | ||
GenBank mRNA | CR406058.1 | - | 2 - 3 | 188 | 72 | |
CR406252.1 | - | 3 - 4 | 282 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |