Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276340.2 | XP_015131826.1 | 26 - 27 | 5424 | 104 | |
XM_015276342.2 | XP_015131828.1 | 26 - 27 | 5382 | 104 | ||
GenBank mRNA | CR406058.1 | - | 5 - 6 | 771 | 104 | |
CR406252.1 | - | 6 - 7 | 865 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |