Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199418.3 | NP_001186347.3 | 6 - 7 | 741 | 80 | |
GenBank mRNA | CR523356.1 | - | 5 - 6 | 633 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |