Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167703.1 | NP_001161175.1 | 3 - 4 | 459 | 91 | |
NM_001167704.1 | NP_001161176.1 | 3 - 4 | 459 | 91 | ||
GenBank mRNA | X56647.1 | - | 459 | 91 | ||
X56648.1 | - | 162 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |