Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282217.1 | NP_001269146.1 | 2 - 3 | 248 | 72 | |
GenBank mRNA | AF043543.1 | - | 1 - 2 | 143 | 72 | |
BG642102.1 | - | 2 - 3 | 240 | 72 | ||
BG713316.1 | - | 2 - 3 | 248 | 72 | ||
BG713347.1 | - | 2 - 3 | 232 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |