Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP7B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147742.1 | XP_025003510.1 | 3 - 4 | 1289 | 97 | |
GenBank mRNA | AY700579.1 | - | 3 - 4 | 813 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |