Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936566.3 | XP_004936623.1 | 4 - 5 | 532 | 132 | |
XM_015290740.2 | XP_015146226.1 | 5 - 6 | 1903 | 132 | ||
XM_015290741.2 | XP_015146227.1 | 4 - 5 | 613 | 132 | ||
XM_422319.5 | XP_422319.2 | 4 - 5 | 1668 | 132 | ||
GenBank mRNA | BX950809.2 | - | 3 - 4 | 352 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |