Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277922.1 | NP_001264851.1 | 6 - 7 | 405 | 149 | |
GenBank mRNA | BU278047.1 | - | 6 - 7 | 270 | 149 | |
CR390466.1 | - | 6 - 8 | 313 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |