Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935234.3 | XP_004935291.1 | 2 - 3 | 794 | 138 | |
XM_015283526.2 | XP_015139012.1 | 4 - 5 | 552 | 138 | ||
XM_015283527.2 | XP_015139013.1 | 4 - 5 | 551 | 138 | ||
XM_015283528.2 | XP_015139014.1 | 3 - 4 | 420 | 138 | ||
XM_025148918.1 | XP_025004686.1 | 5 - 6 | 616 | 138 | ||
XM_025148919.1 | XP_025004687.1 | 3 - 4 | 3415 | 138 | ||
GenBank mRNA | CR405917.1 | - | 1 - 2 | 96 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |