Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416593.6 | XP_416593.1 | 5 - 6 | 762 | 98 | |
GenBank mRNA | BX935082.2 | - | 5 - 6 | 732 | 98 | |
BX950267.2 | - | 5 - 6 | 710 | 98 | ||
CR406044.1 | - | 5 - 6 | 728 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |