Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPTIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277806.1 | NP_001264735.1 | 6 - 7 | 826 | 75 | |
XM_015294031.2 | XP_015149517.1 | 6 - 7 | 838 | 75 | ||
XM_025154866.1 | XP_025010634.1 | 6 - 7 | 832 | 75 | ||
XM_025154867.1 | XP_025010635.1 | 6 - 7 | 843 | 75 | ||
GenBank mRNA | CN219327.1 | - | 2 - 3 | 144 | 75 | |
CR406471.1 | - | 3 - 4 | 658 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |