Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419584.6 | XP_419584.3 | 1 - 2 | 76 | 69 | |
GenBank mRNA | BX950558.1 | - | 1 - 2 | 47 | 69 | |
CR407474.1 | - | 1 - 2 | 87 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |