Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421702.4 | XP_421702.4 | 3 - 4 | 617 | 61 | |
GenBank mRNA | CR389252.1 | - | 2 - 3 | 161 | 61 | |
CR391603.1 | - | 1 - 2 | 122 | 61 | ||
CR523016.1 | - | 2 - 3 | 128 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |