Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299728.2 | XP_015155214.1 | 4 - 5 | 602 | 61 | |
XM_015299729.2 | XP_015155215.1 | 4 - 5 | 604 | 61 | ||
XM_015299730.2 | XP_015155216.1 | 4 - 5 | 603 | 61 | ||
XM_025144422.1 | XP_025000190.1 | 3 - 4 | 197 | 61 | ||
GenBank mRNA | EF520346.1 | - | 2 - 3 | 181 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |