Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167718.1 | NP_001161190.1 | 6 - 7 | 1227 | 71 | |
NM_001167719.1 | NP_001161191.1 | 6 - 7 | 798 | 71 | ||
GenBank mRNA | M64990.1 | - | 6 - 7 | 1323 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |