Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_425129.5 | XP_425129.2 | 3 | 1583 | 82 | |
GenBank mRNA | U62140.1 | - | 1 | 154 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |