Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113708.1 | NP_001107180.1 | 3 - 4 | 295 | 57 | |
NM_001271964.1 | NP_001258893.1 | 3 - 4 | 364 | 57 | ||
XM_025150612.1 | XP_025006380.1 | 10 - 11 | 2927 | 57 | ||
GenBank mRNA | BM490287.1 | - | 3 - 4 | 286 | 57 | |
BU335410.1 | - | 3 - 4 | 260 | 57 | ||
EU367492.1 | - | 2 - 3 | 221 | 57 | ||
X03012.1 | - | 188 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |