Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293098.2 | NP_001280027.1 | 3 - 4 | 760 | 77 | |
GenBank mRNA | X79695.1 | - | 358 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |