Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418776.5 | XP_418776.4 | 3 - 4 | 847 | 95 | |
GenBank mRNA | BX929890.1 | - | 3 - 4 | 432 | 95 |
Gene Symbol: NOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147213.1 | XP_025002981.1 | 3 - 4 | 270 | 95 | |
XM_025147214.1 | XP_025002982.1 | 3 - 4 | 270 | 95 | ||
XM_025147215.1 | XP_025002983.1 | 2 - 3 | 283 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |