Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015285700.2 | XP_015141186.2 | 2 - 3 | 405 | 107 | |
GenBank mRNA | BX929804.2 | - | 1 - 2 | 103 | 107 | |
BX930109.1 | - | 1 - 2 | 90 | 107 | ||
BX930609.1 | - | 1 - 2 | 97 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |