Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277918.1 | NP_001264847.1 | 1 - 2 | 186 | 126 | |
GenBank mRNA | BU264806.1 | - | 1 - 2 | 115 | 126 | |
BX930160.1 | - | 1 - 2 | 94 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |