Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277918.1 | NP_001264847.1 | 5 - 6 | 624 | 80 | |
GenBank mRNA | BU264806.1 | - | 5 - 6 | 553 | 80 | |
BX930160.1 | - | 5 - 6 | 532 | 80 | ||
BX933062.1 | - | 5 - 6 | 494 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |