Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036657.2 | 2 - 3 | 578 | 68 | ||
GenBank mRNA | BX930203.1 | - | 2 - 3 | 200 | 68 | |
BX930271.1 | - | 2 - 3 | 578 | 68 | ||
BX932942.1 | - | 1 - 2 | 214 | 68 | ||
BX934307.2 | - | 2 - 3 | 93 | 68 | ||
BX934976.2 | - | 2 - 3 | 587 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |