Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422088.5 | XP_422088.1 | 3 - 4 | 385 | 93 | |
GenBank mRNA | BX930757.1 | - | 3 - 4 | 364 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |