Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415873.6 | XP_415873.1 | 5 - 6 | 492 | 151 | |
GenBank mRNA | BX929961.1 | - | 4 - 5 | 724 | 151 | |
BX930350.1 | - | 5 - 6 | 442 | 151 | ||
BX930849.1 | - | 5 - 6 | 427 | 151 | ||
BX933199.1 | - | 5 - 6 | 430 | 151 | ||
BX933399.2 | - | 4 - 5 | 473 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |