Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278016.1 | NP_001264945.1 | 2 - 3 | 200 | 81 | |
GenBank mRNA | BU226926.1 | - | 2 - 3 | 186 | 81 | |
BU405971.1 | - | 2 - 3 | 200 | 81 | ||
BX931791.1 | - | 1 - 2 | 214 | 81 | ||
BX950479.2 | - | 2 - 3 | 200 | 81 | ||
CR407359.1 | - | 2 - 3 | 205 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |