Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286368.2 | NP_001273297.1 | 3 - 4 | 393 | 87 | |
NM_001286369.2 | NP_001273298.1 | 2 - 3 | 229 | 87 | ||
XM_015288379.2 | XP_015143865.1 | 4 - 5 | 572 | 87 | ||
XM_015288380.2 | XP_015143866.1 | 4 - 5 | 462 | 87 | ||
XM_015288381.2 | XP_015143867.1 | 4 - 5 | 513 | 87 | ||
XM_015288382.2 | XP_015143868.1 | 4 - 5 | 382 | 87 | ||
XM_015288384.2 | XP_015143870.1 | 3 - 4 | 438 | 87 | ||
XM_025151577.1 | XP_025007345.1 | 5 - 6 | 974 | 87 | ||
XM_025151578.1 | XP_025007346.1 | 5 - 6 | 919 | 87 | ||
XM_025151579.1 | XP_025007347.1 | 4 - 5 | 576 | 87 | ||
XM_025151580.1 | XP_025007348.1 | 4 - 5 | 412 | 87 | ||
XM_025151581.1 | XP_025007349.1 | 3 - 4 | 391 | 87 | ||
XM_025151583.1 | XP_025007351.1 | 3 - 4 | 408 | 87 | ||
XM_025151584.1 | XP_025007352.1 | 3 - 4 | 336 | 87 | ||
GenBank mRNA | BX931503.2 | - | 3 - 4 | 393 | 87 | |
BX931843.1 | - | 2 - 3 | 245 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |