Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277352.1 | NP_001264281.1 | 2 - 3 | 250 | 75 | |
NR_102403.1 | 3 - 4 | 541 | 75 | |||
GenBank mRNA | BU325077.1 | - | 2 - 3 | 250 | 75 | |
BX930539.2 | - | 3 - 4 | 499 | 75 | ||
BX931889.1 | - | 2 - 3 | 243 | 75 | ||
BX932281.2 | - | 2 - 3 | 235 | 75 | ||
BX935089.1 | - | 2 - 3 | 241 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |