Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278110.3 | NP_001265039.1 | 3 - 4 | 120 | ||
GenBank mRNA | BU339707.1 | - | 3 - 4 | 475 | 120 | |
BX932584.1 | - | 3 - 4 | 473 | 120 | ||
BX933150.1 | - | 3 - 4 | 120 | |||
BX933493.2 | - | 3 - 4 | 120 | |||
BX950505.1 | - | 3 - 4 | 405 | 120 | ||
CR386358.1 | - | 3 - 4 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |