Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278112.1 | NP_001265041.1 | 2 - 3 | 175 | 66 | |
GenBank mRNA | BU315783.1 | - | 2 - 3 | 145 | 66 | |
BX930967.1 | - | 2 - 3 | 155 | 66 | ||
BX933167.1 | - | 2 - 3 | 145 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |