Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277399.1 | NP_001264328.1 | 6 - 7 | 579 | 104 | |
GenBank mRNA | BX934448.1 | - | 6 - 7 | 545 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |